Human Recombinant Adiponectin
Catalog # 89151-810
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:50 µg
- Endotoxin Content:<0.1EU/µg purified protein (LAL test; Bio Whittaker).
- Biological Activity:Activates AMPK.
- Reconstitution Instructions:Reconstitute with 50µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum or other carrier protein.
- Protein Synonyms:ACDC
- Protein/Peptide Name:Adiponectin
- Purity:>90% (SDS-PAGE)
- Concentration:1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Cat. No.:89151-810
- Supplier no.:522-063-C050
Specifications
About this item
Produced in HEK 293 cells. Human adiponectin (aa 16-244) is fused at the N-terminus to a linker peptide (14 aa) and a FLAG®-tag.
Mimics serum adiponectin by forming high molecular weight (HMW) and hexameric species.