Human Recombinant RANKL (Soluble)
Catalog # 89151-698
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:10 µg
- Endotoxin Content:<0.1EU/µg purified protein (LAL test; Bio Whittaker).
- Biological Activity:Supports the survival of dendritic cells and osteoclasts.
- Reconstitution Instructions:Reconstitute with 100µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/Peptide Name:RANKL (soluble)
- Purity:>95% (SDS-PAGE)
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Cat. No.:89151-698
- Supplier no.:522-012-C010
Specifications
About this item
Produced in HEK 293 cells. The extracellular domain of human RANKL (aa 152-317) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG®-tag.