Order Entry
Puerto Rico
ContactUsLinkComponent
Adenosine-3’,5’-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (by HPLC)
Adenosine-3’,5’-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (by HPLC)
Catalog # 89151-574
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Adenosine-3’,5’-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (by HPLC)
Catalog # 89151-574
Supplier:  Enzo Life Sciences
Supplier Number:  480-085-M001
CAS Number:  

Specifications

  • Size:
    1 mg
  • Cat. No.:
    89151-574
  • Supplier no.:
    480-085-M001

Specifications

About this item

 :
 :344.3 · 23.0 g/mol
Storage Temperature:Freezer
CAS Number:73208-40-9
Formula:C₁₀H₁₁N₅O₅PS·Na