Order Entry
Puerto Rico
ContactUsLinkComponent
DL-α-Tocopherol (Vitamin E) ≥97%, high purity
DL-α-Tocopherol (Vitamin E) ≥97%, high purity
Catalog # 89151-428
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
DL-α-Tocopherol (Vitamin E) ≥97%, high purity
Catalog # 89151-428
Supplier:  Enzo Life Sciences
Supplier Number:  460-018-G001
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    89151-428
  • Supplier no.:
    460-018-G001

Specifications

About this item

 :
 :430.71 g/mol
CAS Number:10191-41-0
 :1406-66-2
Formula:C₂₉H₅₀O₂
MDL Number:MFCD00072051