Calcium pantothenate ≥98% (by HPLC)
Catalog # 89151-404
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:10 g
- Cat. No.:89151-404
- Supplier no.:460-003-G010
Specifications
Specification Test Results
- Purity (HPLC):≥98%
- Identity:Determined by IR
- Appearance:White to off-white powder
Specification Test Results
About this item
:
:476.54 g/mol
Merck Index:14,07015
Storage Temperature:Ambient
EINECS:205-278-9
CAS Number:137-08-6
Formula:C₁₈H₃₂CaN₂O₁₀
MDL Number:MFCD00002766