Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
D-(+)-Biotin ≥98%

D-(+)-Biotin ≥98%

Supplier:   Enzo Life Sciences
D-(+)-Biotin ≥98%
Supplier:  Enzo Life Sciences
Certificates
D-(+)-Biotin ≥98%
D-(+)-Biotin ≥98%

2 Options Available Below

Product Details & Documents
  • Formulation: White to off-white powder.
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each
    Each - $51.23
    $51.23
    Each
       $51.23
    Minimum order quantity is 1

    Supplier Number: 460-002-G001
  • 5 g
    Log in for availability
    Log in for availability
    Each
    Each - $168.95
    $168.95
    Each
       $168.95
    Minimum order quantity is 1

    Supplier Number: 460-002-G005