Order Entry
Puerto Rico
ContactUsLinkComponent
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98%
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98%
Catalog # 89151-042
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98%
Catalog # 89151-042
Supplier:  Enzo Life Sciences
Supplier Number:  430-081-G001
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    89151-042
  • Supplier no.:
    430-081-G001

Specifications

About this item

 :
 :172.25 g/mol
CAS Number:2226-96-2
Formula:C₉H₁₈NO₂
MDL Number:MFCD00006478