Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Carboxy-PTIO potassium salt
Carboxy-PTIO potassium salt ≥99% (by HPLC)
Carboxy-PTIO potassium salt ≥99% (by HPLC)
Carboxy-PTIO potassium salt ≥99% (by HPLC)
Supplier:
Enzo Life Sciences
Certificates
Expand image
Carboxy-PTIO potassium salt ≥99% (by HPLC)
Supplier:
Enzo Life Sciences
About this item
CAS Number:
148819-94-7
:
Formula:
C₁₄H₁₆KN₂O₄
MDL Number:
MFCD00216153
:
315.4 g/mol
Storage Temperature:
Freezer