Ammonium 1-pyrrolidinecarbodithioate ≥98% (by HPLC)
Catalog # 89150-564
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:5 g
- Cat. No.:89150-564
- Supplier no.:400-002-G005
Specifications
About this item
:
:146.3 · 18.0 g/mol
Storage Temperature:Refrigerator
CAS Number:5108-96-3
Formula:C₅H₁₂N₂S₂
MDL Number:MFCD00012720