Order Entry
Puerto Rico
ContactUsLinkComponent
3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride ≥97% (by HPLC)
3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride ≥97% (by HPLC)
Catalog # 89150-512
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
3,4',5,7-Tetrahydroxy-3'-methoxy flavylium chloride ≥97% (by HPLC)
Catalog # 89150-512
Supplier:  Enzo Life Sciences
Supplier Number:  385-015-M005
CAS Number:  

Specifications

  • Size:
    5 mg
  • Cat. No.:
    89150-512
  • Supplier no.:
    385-015-M005

Specifications

Specification Test Results

  • Purity:
    ≥97% (HPLC)

Specification Test Results

About this item

 :
 :336.73 g/mol
Merck Index:14,07143
Storage Temperature:Refrigerator
EINECS:205-125-6
CAS Number:134-01-0
Formula:C₁₆H₁₃O₆Cl
MDL Number:MFCD00017587