Order Entry
Puerto Rico
ContactUsLinkComponent
Piericidin A (from Streptomyces sp. MST-AS5364) ≥95% (by HPLC)
Piericidin A (from Streptomyces sp. MST-AS5364) ≥95% (by HPLC)
Catalog # 89150-258
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Piericidin A (from Streptomyces sp. MST-AS5364) ≥95% (by HPLC)
Catalog # 89150-258
Supplier:  Enzo Life Sciences
Supplier Number:  380-235-M002
CAS Number:  

Specifications

  • Size:
    2 mg
  • Cat. No.:
    89150-258
  • Supplier no.:
    380-235-M002

Specifications

About this item

 :
 :415.6 g/mol
Storage Temperature:Freezer
CAS Number:2738-64-9
Formula:C₂₅H₃₇NO₄
MDL Number:MFCD01677956