Hispolon ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
:173933-40-9
Formula:C₁₂H₁₂O₄
:220.2 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (HPLC)
- Identity:Determined by IR, MS and NMR
- Source:Synthetic