Order Entry
Puerto Rico
ContactUsLinkComponent
Pyripyropene A ≥85% (by HPLC)
Pyripyropene A ≥85% (by HPLC)
Catalog # 89149-144
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Pyripyropene A ≥85% (by HPLC)
Catalog # 89149-144
Supplier:  Enzo Life Sciences
Supplier Number:  350-134-C500
CAS Number:  

Specifications

  • Size:
    500 µg
  • Cat. No.:
    89149-144
  • Supplier no.:
    350-134-C500

Specifications

About this item

 :
 :583.2 g/mol
CAS Number:147444-03-9
MDL Number:MFCD00929109