Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Helenalin
Helenalin ≥97% (by HPLC)
Helenalin ≥97% (by HPLC)
Helenalin ≥97% (by HPLC)
Catalog # 89149-104
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Helenalin ≥97% (by HPLC)
Catalog # 89149-104
Supplier:
Enzo Life Sciences
Supplier Number:
350-120-MC05
CAS Number:
Specifications
Size:
0.5 mg
Cat. No.:
89149-104
Supplier no.:
350-120-MC05
Specifications
About this item
Product Details
:
CAS Number:
6754-13-8
Product Details