SC-236 ≥95% (by HPLC)
Catalog # 89147-736
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:25 mg
- Cat. No.:89147-736
- Supplier no.:270-377-M025
Specifications
About this item
:
:401.8 g/mol
Storage Temperature:Freezer
CAS Number:170569-86-5
Formula:C₁₆H₁₁ClF₃N₃O₂S
MDL Number:MFCD00941297