Sodium butyrate ≥98.5% (by GC)
Catalog # 89147-604
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:1 g
- Cat. No.:89147-604
- Supplier no.:270-301-G001
Specifications
About this item
:
:110.09 g/mol
CAS Number:156-54-7
Formula:C₄H₇NaO₂
MDL Number:MFCD00002816