To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:TNF Receptor-associated Factor 5
- Antigen Symbol:TRAF5
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:50 µL
- Environmentally Preferable:
- Form:Liquid. In PBS containing 0.05% sodium azide.
- Antigen Synonyms:RNF84|MGC:39780
- Cat. No.:89145-144
- Supplier no.:210-027-R050
Specifications
About this item
Host: Rabbit
- For Immunohistochemistry, Western Blot
Recognizes human and mouse TRAF5. Detects a band of ~64kDa by Western blot. Does not cross-react with other members of the TRAF family.
Type: Primary
Antigen: TRAF5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human