Human Recombinant Control:Fc fusion protein
Catalog # 89145-044
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:50 µg
- Reconstitution Instructions:Reconstitute with 50µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/Peptide Name:Control:Fc fusion protein
- Purity:>90% (SDS-PAGE)
- Concentration:1mg/ml (after reconstitution)
- Formulation:Lyophilized. Contains PBS.
- Cat. No.:89145-044
- Supplier no.:203-004-C050
Specifications
About this item
Negative control for Fc chimeric proteins
Produced in HEK 293 cells. The extracellular domain of mutated hThy-1, hThy-1 (RLE) (aa 20-130), is fused to the Fc portion of human IgG1. The integrin binding sequence RLD of hThy-1 has been changed to RLE abolishing the binding of integrins.
Negative control for Fc chimeric proteins. Does not bind to a series of ligands in an ELISA assay. Does not inhibit binding between the thymoma cell line EL-4 expressing β3-integrin and astrocytes expressing high level of Thy-1 in mice.