Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant HSP40/Hdj2 (from E. coli)
 
undefined
Human Recombinant HSP40/Hdj2 (from E. coli)

 

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Form:
    Liquid
  • Protein/Peptide Name:
    HSP40/Hdj2
  • Purity:
    >90% (SDS-PAGE; Western blot)
  • Formulation:
    In TBS containing 1mM DTT.
  • Purification:
    Purified by multi-step chromatography.