ContactUsLinkComponent
Akt kinase activity kit, Enzo Life Sciences
Akt kinase activity kit, Enzo Life Sciences
Catalog # 89141-836
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Akt kinase activity kit, Enzo Life Sciences
Catalog # 89141-836
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-EKS-400A
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Size:
    96 wells
  • Cat. No.:
    89141-836
  • Supplier no.:
    EKS-400A
Specifications
About this item
For the measurement of Akt in partially purified, purified, or crude enzyme preparations from any species.
This is a non-radioactive Akt kinase activity assay, providing a safe, rapid and reliable method for the screening of inhibitors or activators of Akt and for quantitating the activity of Akt in purified or partially purified enzyme preparations. The Akt Kinase activity kit is based on a solid phase enzyme immunoassay (EIA) that utilizes a specific synthetic peptide as a substrate for Akt and a polyclonal antibody that recognizes the phosphorylated form of the substrate. The assay is designed for the analysis of Akt (1, 2, 3) activity in the solution phase.
Ordering information: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2