Order Entry
Puerto Rico
ContactUsLinkComponent
Catalase Fluorometric Detection Kit, Enzo Life Sciences
Catalase Fluorometric Detection Kit, Enzo Life Sciences
Catalog # 89141-680
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Catalase Fluorometric Detection Kit, Enzo Life Sciences
Catalog # 89141-680
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-907-027
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Catalase fluorometric detection kit
  • Size:
    500 tests
  • Storage Conditions:
    +4 °C (long term)
  • Species Reactivity:
    Species independent
  • Sensitivity:
    0- 4 U/mL
  • Cat. No.:
    89141-680
  • Application:
    Fluorescent detection, HTS
  • Supplier no.:
    907-027

Specifications

About this item

The Catalase fluorometric detection kit provides a higher throughput sensitive method for measuring catalase activity in just one hour

  • Easily adaptable to HTS applications
  • Sensitive fluorescent detection system

For the measurement of catalase in cell lysates from any species.

This is a homogeneous mix-and-read assay that enables you to monitor multiple time points for a kinetic read. Positive control is provided.

Delivery information: Reaction buffer concentrate, Detection Reagent, Horseradish peroxidase concentrate, Hydrogen peroxide, Catalase enzyme