NitraVer® 6 Nitrate Reagent Powder Pillows, 30 ml, Hach
Catalog # 89230-956
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Chemical method:Cadmium reduction
- Description:NitraVer® 6 Nitrate Reagent Powder Pillows
- Size:30 mL
- Method:Cadmium Reduction
- Cat. No.:89230-956
- Range:up to 0.50 mg/L as nitrate-N
- Supplier no.:1411999
- No. of tests:100
Specifications
About this item
For determination of low range nitrate by the cadmium reduction method
Range: up to 0.50 mg/L as nitrate-N. Use with NitriVer® 3. Sample Size: 30 mL.
Ordering information: 100/pk.