TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Catalog # Q-1665.0025BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:25 g
- Cat. No.:Q-1665.0025BA
- Supplier no.:Q-1665.0025
Specifications
About this item
:
:321.1 g/mol
Storage Temperature:Refrigerator
CAS Number:125700-67-6
Formula:C₁₁H₁₆BF₄N₅O
MDL Number:MFCD00077413