Abz-Ser-Pro-3-nitro-Tyr-OH
Selective substrate for angiotensin-converting enzyme 2 (ACE2), an ACE homolog which differs in its specificity and physiological role from ACE. The internally quenched fluorescent (FRET) substrate is potentially useful in applications such as high-throughput screening of ACE2 inhibitors (Km = 23 µM, kcat/Km = 3.5 · 10⁴ M⁻¹s⁻¹).
- CAS No.:553644-01-2
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD09037414
- Protein/Peptide Name:Abz-Ser-Pro-3-nitro-Tyr-OH
- Molecular Weight:529.51
- Sequence:Abz-Ser-Pro-3-nitro-Tyr-OH
- Molecular Formula:C₂₄H₂₇N₅O₉