N-Boc-L-alanine 4-nitrophenyl ester
Catalog # M-1165.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:5 g
- Cat. No.:M-1165.0005BA
- Supplier no.:M-1165.0005
Specifications
About this item
:
:310.306 g/mol
Storage Temperature:Freezer
EINECS:219-621-5
CAS Number:2483-49-0
Formula:C₁₄H₁₈N₂O₆
MDL Number:MFCD00038117