DL-Methionine-β-naphthylamide hydrochloride
Catalog # K-1435.1000BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:1 g
- Cat. No.:K-1435.1000BA
- Supplier no.:K-1435.1000
Specifications
About this item
:
:310.84 g/mol
Storage Temperature:Ambient
CAS Number:97405-58-8
Formula:C₁₅H₁₉ClN₂OS
MDL Number:MFCD00058049