DL-Alanine-β-naphthylamide hydrochloride
Catalog # K-1030.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:1 g
- Cat. No.:K-1030.0001BA
- Supplier no.:K-1030.0001
Specifications
About this item
:
:250.726 g/mol
Storage Temperature:Ambient
EINECS:277-736-6
CAS Number:74144-49-3
Formula:C₁₃H₁₅ClN₂O
MDL Number:MFCD00012563