Bz-Arg-Gly-Phe-Phe-Leu-4MbetaNA Trifluoroacetate
J-1060.0050BA
:
- CAS No.:99112-24-0
- Source:
- Species:
- Size:50 mg
- Environmentally Preferable:
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD03788112
- Protein/Peptide Name:Bz-Arg-Gly-Phe-Phe-Leu-4MbetaNA Trifluoroacetate
- Molecular Weight:898.08
- Sequence:Bz-Arg-Gly-Phe-Phe-Leu-4MβNA
- Molecular Formula:C₅₀H₅₉N₉O₇
- Cat. No.:J-1060.0050BA
- Supplier no.:J-1060.0050
Bz-RGFFL-MNA, substrate for cathepsin D.