Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ac-LEHD-AFC

Ac-LEHD-AFC

5 mg
Catalog # I-1820.0005BA
Supplier:   Bachem Americas
Supplier #:  4028699.0005
Ac-LEHD-AFC
Catalog # I-1820.0005BA
Supplier:  Bachem Americas
CAS Number:  210345-03-2
Ac-LEHD-AFC

5 mg

CAS Number:  210345-03-2
Price
$416.28
Log in for availability
Each (5mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sequence: Ac-Leu-Glu-His-Asp-AFC
Synonym(s): Ac-LEHD-AFC#Caspase 9 Substrate 1f, fluorogenic

Synonyms:  Ac-Leu-Glu-His-Asp-AFC, N-Acetyl-Leu-Glu-His-Asp-7-amino-4-(trifluoromethyl)coumarin
  • CAS Number:210345-03-2
  • Formula:C₃₃H₃₈F₃N₇O₁₁
  • MDL Number:MFCD01862610
  • Molecular Weight:765.7 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Size:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.