Z-Phe-Arg-AMC · HCl
Supplier: Bachem Americas
Certificates
About this item
A fluorogenic substrate for cruzipain, plasma kallikrein and soybean trypsin-like enzyme. Z-FR-AMC was also found to be an excellent substrate for the fluorometric assay of cathepsin B, cathepsin K, and cathepsin L as well as papain. Z-FR-AMC-is also an often-used substrate for falcipain I and II.
Specifications
- Source:
- Species:
- Environmentally Preferable: