Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Iodo-L-tyrosine
3-Iodo-L-tyrosine
3-Iodo-L-tyrosine
3-Iodo-L-tyrosine
Supplier:
Bachem Americas
Certificates
Expand image
3-Iodo-L-tyrosine
Supplier:
Bachem Americas
About this item
CAS Number:
70-78-0
:
Formula:
C₉H₁₀INO₃
MDL Number:
MFCD00002608
:
307.1 g/mol
Storage Temperature:
Ambient
EINECS:
200-744-8