S-Methyl-L-thiocitrulline
Catalog # F-3180.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:50 mg
- Cat. No.:F-3180.0050BA
- Supplier no.:F-3180.0050
Specifications
About this item
:
:205.28 g/mol
Storage Temperature:Freezer
CAS Number:156719-41-4
Formula:C₇H₁₅N₃O₂S
MDL Number:MFCD00672722