H-DL-allo-Ile-OH
Catalog # F-1170.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:1 g
- Cat. No.:F-1170.0001BA
- Supplier no.:F-1170.0001
Specifications
About this item
:
:131.18 g/mol
Storage Temperature:Ambient
EINECS:221-464-2
CAS Number:3107-04-8
Formula:C₆H₁₃NO₂
MDL Number:MFCD00065934