Ac-P-Amino-Phe-Ome
Catalog # F-1015.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:1 g
- Cat. No.:F-1015.0001BA
- Supplier no.:F-1015.0001
Specifications
About this item
:
:236.271 g/mol
Storage Temperature:Freezer
CAS Number:36097-42-4
Formula:C₁₂H₁₆N₂O₃
MDL Number:MFCD00025378