L-Leucinamide hydrochloride
Catalog # E-2055.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:5 g
- Cat. No.:E-2055.0005BA
- Supplier no.:E-2055.0005
Specifications
About this item
:
:166.65 g/mol
Storage Temperature:Refrigerator
EINECS:233-952-2
CAS Number:10466-61-2
Formula:C₆H₁₄N₂O
MDL Number:MFCD00013012