Z-L-Thr-OSu
Catalog # C-2650.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:5 g
- Cat. No.:C-2650.0005BA
- Supplier no.:C-2650.0005
Specifications
About this item
:
:350.327 g/mol
Storage Temperature:Freezer
CAS Number:76401-90-6
Formula:C₁₆H₁₈N₂O₇
MDL Number:MFCD00153344