Order Entry
Puerto Rico
ContactUsLinkComponent
 
Boc-Ser(Leu-Fmoc)-OH
 
Boc-Ser(Leu-Fmoc)-OH

 

  • CAS No.:
    944283-09-4
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD11974990
  • Protein/Peptide Name:
    Boc-Ser(Leu-Fmoc)-OH
  • Molecular Weight:
    540.61
  • Sequence:
    Boc-Ser(Leu-Fmoc)-OH
  • Molecular Formula:
    C₂₉H₃₆N₂O₈