Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Toolbox fmoc-amino acid derivatives
Toolbox fmoc-amino acid derivatives
Toolbox fmoc-amino acid derivatives
Toolbox fmoc-amino acid derivatives
Catalog # B-3840.0001BA
Supplier:
Bachem Americas
CAS Number:
Expand image
Toolbox fmoc-amino acid derivatives
Catalog # B-3840.0001BA
Supplier:
Bachem Americas
Supplier Number:
4050008.0001
CAS Number:
Specifications
Size:
100 g
Cat. No.:
B-3840.0001BA
Supplier no.:
B-3840.0001
Specifications
About this item
Product Details
:
Storage Temperature:
Freezer
Product Details