(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((2,4-dinitrophenyl)amino)propanoic acid
Catalog # B-2995.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:1 g
- Cat. No.:B-2995.0001BA
- Supplier no.:B-2995.0001
Specifications
About this item
:
:492.444 g/mol
Storage Temperature:Freezer
CAS Number:140430-54-2
Formula:C₂₄H₂₀N₄O₈
MDL Number:MFCD00798650