Fmoc-Ser(tBu)-ODhbt
Catalog # B-1640.0025BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:25 g
- Cat. No.:B-1640.0025BA
- Supplier no.:B-1640.0025
Specifications
About this item
:
:528.565 g/mol
Storage Temperature:Freezer
CAS Number:109434-27-7
Formula:C₂₉H₂₈N₄O₆
MDL Number:MFCD00133600