Fmoc-Phe-OSu
Catalog # B-1415.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:5 g
- Cat. No.:B-1415.0005BA
- Supplier no.:B-1415.0005
Specifications
About this item
:
:484.508 g/mol
Storage Temperature:Freezer
CAS Number:101214-43-1
Formula:C₂₈H₂₄N₂O₆
MDL Number:MFCD00062204