Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-L-Arg(Mtr)-OH
Fmoc-L-Arg(Mtr)-OH
Fmoc-L-Arg(Mtr)-OH
Fmoc-L-Arg(Mtr)-OH
Supplier:
Bachem Americas
Certificates
Expand image
Fmoc-L-Arg(Mtr)-OH
Supplier:
Bachem Americas
About this item
CAS Number:
98930-01-9
:
Formula:
C₃₁H₃₆N₄O₇S
MDL Number:
MFCD00043098
:
608.71 g/mol
Storage Temperature:
Freezer