$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Fmoc-Ser(tBu)-OPfp
Fmoc-Ser(tBu)-OPfp
Fmoc-Ser(tBu)-OPfp
Fmoc-Ser(tBu)-OPfp
Supplier:
Bachem Americas
Certificates
Expand image
Fmoc-Ser(tBu)-OPfp
Supplier:
Bachem Americas
About this item
CAS Number:
105751-13-1
:
Formula:
C₂₈H₂₄F₅NO₅
MDL Number:
MFCD00062213
:
549.494 g/mol
Storage Temperature:
Freezer