N-Fmoc-L-alanine, hydrate
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-(9-Fluorenylmethoxycarbonyl)-L-alanine, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine, Fmoc-L-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine, (-)-L-Alanine, N-FMOC protected, (-)-FMOC-L-Ala-OH, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected, Fmoc-Ala-OH
CAS Number:35661-39-3
:
Formula:C₁₈H₁₇NO₄
MDL Number:MFCD00037139
Molecular Weight:311.34 g/mol (anhy)
Storage Temperature:Ambient
EINECS:252-660-6
Specifications
- Environmentally Preferable: