Distillation Receiver, Angled Upper Joint, Ace Glass
Distillation Receiver
Catalog # 89074-452
Supplier: Ace Glass
Supplier #: 9389-02
Jump to:
Product Details & Documents
This receiver has an upper joint at a 105° angle
It is designed for operation other than at atmospheric pressure. Vacuum is applied to the side arm with the T-bore stopcock. The secondary receiver is vented by means of the lower stopcock. The capacity from the bottom of the drip to the stopcock is 2.5 mL. [ST] 14/20 outer joint at top; [ST] 14/20 inner joint at bottom. Tubing connections are 8 mm O.D. olive type. Fabricated from borosilicate glass.
Specifications
- Description:Distillation Receiver
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



