Corning® BioCoat™ Control Cell Culture Inserts, PET Membrane, Corning
: Corning
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel® matrix, fibronectin, collagen and laminin.
- Catalog No:
- 62405-000
- 62405-016
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- ISO 14001 - Bedford (reg. no. 14.4942)
- ISO 14001 - Bedford (reg. no. 14.4942)