Order Entry
Puerto Rico
ContactUsLinkComponent
Fixed Dry Block Heater, BTD, Grant Instruments
Fixed Dry Block Heater, BTD, Grant Instruments
Catalog # 97011-930
Supplier:  GRANT USA INC.
undefined
Fixed Dry Block Heater, BTD, Grant Instruments
Catalog # 97011-930
Supplier:  GRANT USA INC.
Supplier Number:  BTD USA

Specifications

  • Capacity:
    24× 1.5/2.0ml, 15× 0.5ml, 10× 0.2ml
  • Description:
    BTD Digital Dry Block Heater for Microtubes
  • Safety Over-Temperature Cut-Out:
    Thermal Fuse
  • Heating Time:
    25 to 100°C in 15 mins.
  • Cat. No.:
    97011-930
  • W×D×H:
    210 x 230 x 110 mm
  • Electrical:
    120 V, 50/60 Hz, 200 W
  • Weight:
    2.5 kg (5.5 lbs.)
  • Supplier no.:
    BTD USA

Specifications

About this item

Compact digital block heater with fixed circular block

  • Stability and uniformity ±0.1ºC
  • Digital temperature control for optimum precision
  • Temperature range: ambient +5ºC to 100ºC, with rapid heat-up time
  • Capacity for up to 49 microtubes in a combination of four common sizes
  • Integral timer

The aluminium block is housed in durable and easy to clean, chemically resistant plastic. Unit has a 0.1°C display resolution. The BTD is ideal for use in DNA, PCR and ELISA applications as well as for General heating samples.

Unit features digital control, two line display allowing simple and accurate setting of temperature and time, plus display of current status while in operation

Certifications: CE approved.
Ordering information: System includes manufacturer's two-year warranty. A 230V version is available. Contact your Avantor representative for additional information.