About this item
Criterion™ Tris-HCl Precast Protein Gels, midi gels, for vertical electrophoresis are compatible with both denaturing and nondenaturing separations
Numbered, marked well outlines on the cassette for easy sample loading and identification
Without SDS-containing buffers, proteins are separated by mass-to-charge ratio
Fast setup, loading, and run times
Run 2-D gels in less than a day
Under denaturing conditions using standard Tris/glycine/SDS buffers, Tris-HCl gels have Laemmli-like separation patterns. Criterion™ Tris-HCl Gels provide consistent and accurate molecular weight estimation in both 1-D and 2-D gel protein separations.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 101645-804
- 101645-808
- 101645-900
- 101645-784
- 101645-780
- 101645-788
- 101645-748
- 101645-806
- 101645-890
- 101645-764
- 101645-758
- 101645-756
- 101645-792
- 101645-760
- 101645-782
- 101645-790
- 101645-960
- 101645-752
- 101645-750
- 101645-768
- 101645-766
- 101645-894
- 101645-888
- Cassette Dimensions:
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- Cassette Material:
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Comb Material:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Format:
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Gel Dimensions:
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- Shelf Life:
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- 12 weeks from date of manufacture
- Storage Conditions:
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Store flat at 2–8°C; do not freeze
- Tray Material:
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET