Order Entry
Puerto Rico
ContactUsLinkComponent
Remote RH/Temperature Monitors, Sper Scientific
Remote RH/Temperature Monitors, Sper Scientific
Catalog # 97035-778
Supplier:  Sper Scientific
undefined
Remote RH/Temperature Monitors, Sper Scientific
Catalog # 97035-778
Supplier:  Sper Scientific
Supplier Number:  800027

Specifications

  • Description:
    Remote RH/Temperature Monitor
  • Dimensions:
    110 x 97 x 20 mm (4.25 x 3.75 x .875”)
  • Weight:
    6 oz (170 g)
  • Cat. No.:
    97035-778
  • Supplier no.:
    800027

Specifications

About this item

Remote relative humidity monitor and digital thermometer.

The remote ambient temperature/relative humidity sensor is mounted on a cord over 7’ long. Place the highly accurate sensor inside a desiccator, refrigerator, drying or other environmental chamber and view simultaneous displays of temperature and RH outside on the large LCD. Recalls min/max readings on both parameters. ºC and ºF selectable. Stands or wall mounts.

Certifications: N.I.S.T. traceable certificate of compliance (for certified model only).
Ordering information: Five year warranty.