Orders LinkContactUsLinkComponent
iScript RT-qPCR Sample Preparation Reagent, Bio-Rad
iScript RT-qPCR Sample Preparation Reagent, Bio-Rad
Marketsource
Supplier:   Bio-Rad
iScript RT-qPCR Sample Preparation Reagent, Bio-Rad
Supplier:  Bio-Rad
Marketsource
Certificates
iScript RT-qPCR Sample Preparation Reagent, Bio-Rad
iScript RT-qPCR Sample Preparation Reagent, Bio-Rad
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
iScript™ RT-qPCR sample preparation reagent provides a simple and rapid procedure (<10 min) for isolating total RNA from cultured cells to use for RT-qPCR. This reagent delivers efficient cell lysis, RNA stabilization, and removal of genomic DNA for sensitive qPCR without traditional RNA sample preparation.
* Have ease of use and simplicity — one-buffer formulation contains all of the necessary components to perform cell lysis, RNA stabilization, and genomic DNA removal in 5–10 min
Key Features and Benefits:
* Have ease of use and simplicity — one-buffer formulation contains all of the necessary components to perform cell lysis, RNA stabilization, and genomic DNA removal in 5–10 min
* Benefit from superior compatibility with qPCR directly from cells — sensitive detection of high-, medium-, and low-copy gene targets directly from cell lysates using real-time qPCR when combined with iScript reverse transcription and qPCR reagents
* Maximize data from very few cells — reagent enables multiplex real-time detection of up to 4 targets from as few as 10 cells
* Extend isolated RNA shelf-life — isolated RNA is stable for up to 6 months when frozen
* Use with different cell types — compatible with both suspension and adherent cell cultures
Packaging: Contains RNase inhibitors and RNA stabilizers
Documents
Product Family Options
Product Information
  • Size
    Volume
    Availability
    Price
  • 101645-420
    iScript RT-qPCR Sample Preparation Reagent
    iScript RT-qPCR Sample Preparation Reagent
    500 reactions
    5× 10 ml
    Log in for availability
    Log in for availability
    Each
    Each - $1,221.54
    $1,221.54
    Each
       $1,221.54
    Supplier Number: 1708899
    Specifications:
    Marketsource
    Marketsource
  • 101645-418
    iScript RT-qPCR Sample Preparation Reagent
    iScript RT-qPCR Sample Preparation Reagent
    100 reactions
    10 ml
    Log in for availability
    Log in for availability
    Each
    Each - $349.23
    $349.23
    Each
       $349.23
    Supplier Number: 1708898
    Specifications:
    Marketsource
    Marketsource